Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF479 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033273.2 | NP_150376.1 | 5 | 1209 | ||
XM_011515604.2 | XP_011513906.1 | 6 | 1395 | |||
XM_011515608.2 | XP_011513910.1 | 6 | 1299 | |||
XM_011515609.2 | XP_011513911.1 | 4 | 1190 | |||
XM_011515610.2 | XP_011513912.1 | 7 | 1476 | |||
XM_011515611.2 | XP_011513913.1 | 5 | 1343 | |||
XM_017012777.1 | XP_016868266.1 | 4 | 1044 | |||
XM_017012778.1 | XP_016868267.1 | 5 | 1247 | |||
GenBank mRNA | AF277624.1 | - | 5 | 1209 |
Gene Symbol: ZNF733P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003952.1 | 3 | 909 |