Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC189 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014979.2 | NP_001014979.2 | 9 | 1430 | ||
XM_011545976.1 | XP_011544278.1 | 8 | 1373 | |||
XM_011545978.1 | XP_011544280.1 | 6 | 1218 | |||
XM_017023852.1 | XP_016879341.1 | 8 | 1658 | |||
XM_017023853.1 | XP_016879342.1 | 7 | 1595 | |||
XM_017023854.1 | XP_016879343.1 | 7 | 1568 | |||
XM_017023855.1 | XP_016879344.1 | 9 | 1562 | |||
XM_017023856.1 | XP_016879345.1 | 8 | 1179 | |||
XM_017023857.1 | XP_016879346.1 | 7 | 1649 | |||
XM_017023858.1 | XP_016879347.1 | 6 | 1498 | |||
XM_017023859.1 | XP_016879348.1 | 7 | 1374 | |||
XM_017023860.1 | XP_016879349.1 | 5 | 833 | |||
XM_017023861.1 | XP_016879350.1 | 8 | 1444 | |||
XM_017023862.1 | XP_016879351.1 | 7 | 1976 | |||
GenBank mRNA | AL122109.2 | - | 6 | 2012 | ||
BC042548.1 | - | 6 | 1645 | |||
BC092491.1 | - | 8 | 1722 | 4837696 | ||
BC128128.1 | - | 8 | 1086 | |||
BC128129.1 | - | 9 | 934 | 40112368 |
Gene Symbol: PHKG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000294.2 | NP_000285.1 | 10 | 1861 | ||
GenBank mRNA | AB209055.1 | - | 5 | 1250 |