Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PKMYT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258450.1 | NP_001245379.1 | 4 | 1071 | ||
NM_001258451.1 | NP_001245380.1 | 4 | 1142 | |||
NM_004203.4 | NP_004194.3 | 4 | 1278 | |||
NM_182687.2 | NP_872629.1 | 4 | 1278 | |||
XM_011522734.2 | XP_011521036.1 | 4 | 1239 | |||
XM_011522735.2 | XP_011521037.1 | 4 | 1065 | |||
XM_011522736.2 | XP_011521038.1 | 3 | 1072 | |||
GenBank mRNA | AK097642.1 | - | 4 | 1027 | ||
AK098452.1 | - | 4 | 1130 | |||
AK294379.1 | - | 3 | 1039 | |||
AK301926.1 | - | 4 | 947 | |||
AK303005.1 | - | 4 | 1156 | |||
AK309964.1 | - | 3 | 1072 | |||
BC121161.1 | - | 3 | 966 | |||
BC121162.1 | - | 3 | 966 | |||
BC130011.1 | - | 3 | 966 | |||
BC153823.1 | - | 3 | 966 | |||
U56816.1 | - | 3 | 982 |