Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033549.4 | NP_291027.3 | 3 | 1691 | ||
NM_201627.2 | NP_963921.1 | 3 | 1691 | |||
NR_045218.1 | 3 | 1691 | ||||
XM_006714929.2 | XP_006714992.1 | 3 | 1689 | |||
XM_006714930.2 | XP_006714993.1 | 3 | 1689 | |||
XM_006714931.3 | XP_006714994.1 | 3 | 1689 | |||
XM_006714932.3 | XP_006714995.1 | 3 | 1689 | |||
GenBank mRNA | AB063180.1 | - | 3 | 717 | ||
AB100366.1 | - | 3 | 954 | |||
AB100367.1 | - | 3 | 954 | |||
AK027601.1 | - | 3 | 1664 | |||
AK074052.1 | - | 2 | 2609 | |||
AK307695.1 | - | 2 | 96 | |||
AL137593.1 | - | 1 | 301 | |||
AL832960.1 | - | 3 | 568 | |||
AX775789.1 | - | 3 | 430 | |||
AX775791.1 | - | 3 | 430 | |||
AX775793.1 | - | 3 | 1664 | |||
AX775797.1 | - | 3 | 722 | |||
AX775799.1 | - | 3 | 1664 | |||
AX775801.1 | - | 3 | 1664 | |||
AX775803.1 | - | 3 | 722 | |||
BC004956.1 | - | 3 | 896 | |||
BC018765.1 | - | 3 | 722 | |||
BC071887.1 | - | 3 | 896 | |||
BC090953.1 | - | 3 | 1234 | 6017628 |