Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243759.1 | NP_001230688.1 | 11 | 1141 | ||
NM_004205.4 | NP_004196.4 | 12 | 1956 | |||
NM_171997.2 | NP_741994.1 | 11 | 1221 | |||
XM_005271721.4 | XP_005271778.1 | 12 | 1829 | |||
XM_005271722.2 | XP_005271779.1 | 12 | 1835 | |||
XM_017018539.1 | XP_016874028.1 | 11 | 917 | |||
GenBank mRNA | AF079564.1 | - | 11 | 959 | ||
AF440755.1 | - | 11 | 1221 | |||
AK057225.1 | - | 12 | 1885 | |||
AK292255.1 | - | 11 | 1144 | |||
AK301805.1 | - | 11 | 1026 | |||
BC002854.1 | - | 12 | 1837 | 3635143 | ||
BC002955.1 | - | 12 | 1837 | 3543435 | ||
BC041366.2 | - | 11 | 1126 | 5273400 |