Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC16A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001201546.1 | NP_001188475.1 | 3 | 422 | ||
NM_001201549.1 | NP_001188478.1 | 3 | 422 | |||
NM_004696.2 | NP_004687.1 | 3 | 422 | |||
XM_005271317.4 | XP_005271374.1 | 4 | 1440 | |||
GenBank mRNA | AK129985.1 | - | 3 | 366 | ||
AK223307.1 | - | 3 | 366 | |||
AK290720.1 | - | 3 | 366 | |||
AK313735.1 | - | 3 | 384 | |||
AL833619.1 | - | 3 | 363 | |||
BC021664.1 | - | 3 | 392 | 4050756 | ||
BP345760.1 | - | 3 | 422 | |||
U59185.1 | - | 3 | 354 |