Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC16A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001201546.1 | NP_001188475.1 | 5 | 909 | ||
NM_001201547.1 | NP_001188476.1 | 5 | 920 | |||
NM_001201548.1 | NP_001188477.1 | 4 | 776 | |||
NM_004696.2 | NP_004687.1 | 6 | 1053 | |||
XM_005271317.4 | XP_005271374.1 | 7 | 2071 | |||
XM_006711033.3 | XP_006711096.1 | 4 | 1662 | |||
GenBank mRNA | AK129985.1 | - | 6 | 997 | ||
AK223307.1 | - | 6 | 997 | |||
AK290720.1 | - | 5 | 853 | |||
AK295946.1 | - | 5 | 886 | |||
AK298539.1 | - | 4 | 720 | |||
AK313735.1 | - | 6 | 1015 | |||
AL833619.1 | - | 5 | 850 | |||
U59185.1 | - | 6 | 985 |