Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC39A13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128225.2 | NP_001121697.1 | 9 | 1151 | ||
NM_152264.4 | NP_689477.2 | 9 | 1151 | |||
NR_134854.1 | 9 | 1216 | ||||
XM_006718381.3 | XP_006718444.1 | 9 | 1035 | |||
XM_006718383.3 | XP_006718446.1 | 8 | 927 | |||
XM_006718384.2 | XP_006718447.1 | 9 | 1031 | |||
XM_006718385.2 | XP_006718448.1 | 8 | 923 | |||
XM_011520466.1 | XP_011518768.1 | 9 | 1056 | |||
XM_011520467.1 | XP_011518769.1 | 9 | 1385 | |||
XM_011520468.2 | XP_011518770.1 | 9 | 1478 | |||
XM_011520469.1 | XP_011518771.1 | 8 | 948 | |||
XM_011520470.1 | XP_011518772.1 | 8 | 922 | |||
XM_017018540.1 | XP_016874029.1 | 9 | 1009 | |||
XM_017018541.1 | XP_016874030.1 | 8 | 901 | |||
XM_017018542.1 | XP_016874031.1 | 9 | 1005 | |||
GenBank mRNA | AK098651.1 | - | 9 | 1216 | ||
AL133581.1 | - | 2 | 123 | |||
BC008853.2 | - | 9 | 1017 | 4100291 | ||
BC019016.2 | - | 9 | 1015 | 4100057 | ||
BX647491.1 | - | 8 | 2504 |