Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BOD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159651.1 | NP_001153123.1 | 2 | 572 | ||
NM_138369.2 | NP_612378.1 | 2 | 572 | |||
GenBank mRNA | AK301982.1 | - | 2 | 560 | ||
AY303777.1 | - | 2 | 345 | |||
BC003114.1 | - | 2 | 497 | 3537973 | ||
BC007436.2 | - | 2 | 562 | 2961368 |