Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF185 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135825.1 | NP_001129297.1 | 2, 3 | 375 | ||
NM_152267.3 | NP_689480.2 | 2, 3 | 375 | |||
NR_024209.1 | 3, 4 | 473 | ||||
NR_024210.1 | 2, 3 | 375 | ||||
NR_024211.1 | 2, 3 | 375 | ||||
GenBank mRNA | AK095947.1 | - | 2, 3 | 284 | ||
AK291236.1 | - | 2, 3 | 284 | |||
AK297414.1 | - | 2, 3 | 345 | |||
BC009504.2 | - | 2, 3 | 324 | 2964524 | ||
BC012817.1 | - | 2, 3 | 333 | 2964524 | ||
BC026040.1 | - | 2, 3 | 267 | 4866294 | ||
BC033166.1 | - | 2, 3 | 310 | 4549320 | ||
BC035684.1 | - | 2, 3 | 334 | 5750180 | ||
BX648019.1 | - | 3, 4 | 397 | |||
CR456349.1 | - | 2, 3 | 263 | |||
DB134817.1 | - | 2, 3 | 375 | |||
DQ296559.1 | - | 2, 3 | 316 | |||
DQ296560.1 | - | 2, 3 | 316 | |||
DQ296561.1 | - | 2, 3 | 316 | |||
DQ296562.1 | - | 2, 3 | 316 | |||
DQ296563.1 | - | 2, 3 | 316 | |||
DQ296564.1 | - | 2, 3 | 316 | |||
DQ296565.1 | - | 2, 3 | 316 |