Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL23A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173465.3 | NP_775736.2 | 2 | 689 | ||
XM_006714933.3 | XP_006714996.1 | 2 | 419 | |||
XM_006714934.3 | XP_006714997.1 | 2 | 419 | |||
XM_006714935.3 | XP_006714998.1 | 2 | 419 | |||
XM_006714936.3 | XP_006714999.1 | 2 | 419 | |||
XM_011534688.2 | XP_011532990.1 | 2 | 419 | |||
XM_011534689.2 | XP_011532991.1 | 2 | 419 | |||
XM_011534690.2 | XP_011532992.1 | 2 | 419 | |||
XM_011534691.2 | XP_011532993.1 | 2 | 419 | |||
XM_011534692.2 | XP_011532994.1 | 2 | 1444 | |||
XM_017010018.1 | XP_016865507.1 | 2 | 419 | |||
GenBank mRNA | AY158895.1 | - | 2 | 689 | ||
AY898961.1 | - | 2 | 689 | |||
BC042428.1 | - | 3 | 395 | 5165083 |