Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF830 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052857.3 | NP_443089.3 | 1 | 858 | ||
GenBank mRNA | AK055707.1 | - | 1 | 842 | ||
AK315671.1 | - | 1 | 858 | |||
BC002913.2 | - | 1 | 803 | |||
BC009044.1 | - | 1 | 421 | |||
BC011584.1 | - | 1 | 803 |