Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NEXN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172309.1 | NP_001165780.1 | 12 | 1982 | ||
NM_144573.3 | NP_653174.3 | 13 | 2174 | |||
XM_005271322.3 | XP_005271379.1 | 13 | 2065 | |||
XM_005271323.3 | XP_005271380.1 | 12 | 2023 | |||
XM_005271324.4 | XP_005271381.1 | 12 | 1969 | |||
XM_005271325.3 | XP_005271382.1 | 12 | 1843 | |||
XM_005271326.3 | XP_005271383.1 | 11 | 1858 | |||
XM_005271327.3 | XP_005271384.1 | 10 | 1648 | |||
GenBank mRNA | AF114264.1 | - | 9 | 1604 | ||
AK057954.1 | - | 9 | 1396 | |||
AK127535.1 | - | 5 | 1451 | |||
AK293201.1 | - | 6 | 1087 | |||
AK298565.1 | - | 12 | 1951 | |||
BC111395.1 | - | 9 | 1563 | 4271572 | ||
BC114444.1 | - | 9 | 1580 | 40083839 | ||
BC114445.1 | - | 9 | 1580 | 40083847 | ||
DQ464902.1 | - | 13 | 2061 |