Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NEXN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172309.1 | NP_001165780.1 | 2, 3 | 334 | ||
NM_144573.3 | NP_653174.3 | 3, 4 | 526 | |||
XM_005271322.3 | XP_005271379.1 | 3, 4 | 417 | |||
XM_005271323.3 | XP_005271380.1 | 3, 4 | 417 | |||
XM_005271324.4 | XP_005271381.1 | 2, 3 | 321 | |||
XM_005271325.3 | XP_005271382.1 | 3, 4 | 417 | |||
XM_005271326.3 | XP_005271383.1 | 2, 3 | 252 | |||
XM_005271327.3 | XP_005271384.1 | 3, 4 | 417 | |||
GenBank mRNA | AF176780.1 | - | 2, 3 | 205 | ||
AK298565.1 | - | 2, 3 | 303 | |||
AK300728.1 | - | 3, 4 | 459 | |||
AK307487.1 | - | 2, 3 | 252 | |||
AY211912.1 | - | 3, 4 | 379 | |||
BC055084.1 | - | 3, 4 | 303 | |||
BX648546.1 | - | 3, 4 | 414 | |||
CR936630.1 | - | 3, 4 | 436 | |||
DC412178.1 | - | 2, 3 | 335 | |||
DQ464902.1 | - | 3, 4 | 413 |