Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF765 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040185.1 | NP_001035275.1 | 4 | 674 | ||
GenBank mRNA | AK293890.1 | - | 6 | 888 | ||
AK316300.1 | - | 4 | 668 | |||
BC140724.1 | - | 4 | 648 | 9021592 | ||
BX647981.1 | - | 4 | 666 |