Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD3E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000733.3 | NP_000724.1 | 5, 6 | 353 | ||
GenBank mRNA | AK292612.1 | - | 5, 6 | 202 | ||
AK301255.1 | - | 5, 6 | 203 | |||
BC049847.1 | - | 4, 5 | 165 | 5214563 | ||
BI910359.1 | - | 5, 6 | 388 | |||
BP366535.1 | - | 5, 6 | 370 | |||
X03884.1 | - | 4, 5 | 151 |