Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IDI2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033261.2 | NP_150286.1 | 3 | 273 | ||
GenBank mRNA | AF271725.1 | - | 2 | 249 | ||
AK056950.1 | - | 3 | 273 | |||
BC017778.1 | - | 3 | 272 | 4291149 |
Gene Symbol: IDI2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024628.1 | 1 | 70 | |||
NR_024629.1 | 1 | 70 | ||||
NR_027708.1 | 1 | 70 | ||||
NR_027709.1 | 1 | 70 | ||||
GenBank mRNA | AF220183.1 | - | 1 | 41 | ||
AK314921.1 | - | 1 | 70 | |||
BF724542.1 | - | 1 | 33 |