Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FDXACB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138378.2 | NP_612387.1 | 5 | 1147 | ||
NR_038364.1 | 4 | 990 | ||||
GenBank mRNA | AK097483.1 | - | 4 | 978 | ||
AK300795.1 | - | 4 | 621 | |||
BC006136.1 | - | 5 | 826 | |||
BC127680.1 | - | 5 | 870 | 40127680 |