Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COG7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153603.3 | NP_705831.1 | 12 | 1692 | ||
XM_017023870.1 | XP_016879359.1 | 12 | 1735 | |||
GenBank mRNA | AF070568.1 | - | 4 | 264 | ||
AK093989.1 | - | 9 | 1048 | |||
AY358632.1 | - | 7 | 729 | |||
BC000549.2 | - | 12 | 1647 | 3163316 | ||
BC037563.1 | - | 12 | 1681 | 5393265 | ||
KU178837.1 | - | 12 | 1495 |