Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COG7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153603.3 | NP_705831.1 | 3 | 611 | ||
XM_017023870.1 | XP_016879359.1 | 3 | 654 | |||
GenBank mRNA | BC000549.2 | - | 3 | 566 | 3163316 | |
BC037563.1 | - | 3 | 600 | 5393265 | ||
KU178837.1 | - | 3 | 414 | |||
KU178838.1 | - | 3 | 414 |