Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC16A7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270622.1 | NP_001257551.1 | 5 | 1371 | ||
NM_001270623.1 | NP_001257552.1 | 5 | 1391 | |||
NM_004731.4 | NP_004722.2 | 4 | 1229 | |||
NR_073055.1 | 6 | 1505 | ||||
NR_073056.1 | 6 | 1366 | ||||
XM_005269231.4 | XP_005269288.1 | 4 | 1165 | |||
XM_011538989.2 | XP_011537291.1 | 6 | 1575 | |||
XM_011538990.2 | XP_011537292.1 | 5 | 1467 | |||
XM_011538991.2 | XP_011537293.1 | 5 | 1265 | |||
XM_011538992.2 | XP_011537294.1 | 5 | 2200 | |||
XM_011538993.2 | XP_011537295.1 | 7 | 1965 | |||
XM_011538995.2 | XP_011537297.1 | 7 | 1960 | |||
XM_017020224.1 | XP_016875713.1 | 8 | 1886 | |||
XM_017020225.1 | XP_016875714.1 | 8 | 1879 | |||
XM_017020226.1 | XP_016875715.1 | 8 | 2014 | |||
XM_017020227.1 | XP_016875716.1 | 7 | 1560 | |||
GenBank mRNA | AF049608.1 | - | 5 | 1226 | ||
AF058056.1 | - | 4 | 1192 | |||
AK301725.1 | - | 6 | 1493 | |||
AK310281.1 | - | 6 | 1333 | |||
AK313345.1 | - | 4 | 1199 | |||
BC030693.2 | - | 4 | 1221 | 4827290 |