Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPGS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033513.2 | NP_277048.2 | 1 | 238 | ||
GenBank mRNA | AK125401.1 | - | 1 | 235 | ||
AK126170.1 | - | 1 | 238 | |||
BC009520.1 | - | 1 | 124 | |||
BM846833.1 | - | 1 | 238 |