Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNX29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032167.4 | NP_115543.3 | 15 | 1827 | ||
XM_005255682.3 | XP_005255739.1 | 15 | 1852 | |||
XM_011522738.2 | XP_011521040.1 | 15 | 1999 | |||
XM_011522741.2 | XP_011521043.1 | 15 | 1983 | |||
XM_011522743.2 | XP_011521045.1 | 15 | 1987 | |||
XM_011522744.2 | XP_011521046.1 | 15 | 1858 | |||
XM_017023871.1 | XP_016879360.1 | 15 | 1996 | |||
XM_017023872.1 | XP_016879361.1 | 15 | 1816 | |||
XM_017023873.1 | XP_016879362.1 | 15 | 1983 | |||
XM_017023874.1 | XP_016879363.1 | 15 | 1816 | |||
XM_017023875.1 | XP_016879364.1 | 13 | 1523 | |||
XM_017023876.1 | XP_016879365.1 | 14 | 1778 | |||
XM_017023878.1 | XP_016879367.1 | 5 | 1321 | |||
GenBank mRNA | AK074072.1 | - | 5 | 434 | ||
BC029857.1 | - | 3 | 221 |