Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZMYM5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039649.2 | NP_001034738.1 | 4 | 827 | ||
NM_001039650.2 | NP_001034739.1 | 4 | 827 | |||
NM_001142684.1 | NP_001136156.1 | 4 | 827 | |||
XM_005266593.3 | XP_005266650.1 | 4 | 1214 | |||
XM_005266594.2 | XP_005266651.1 | 3 | 642 | |||
XM_006719894.2 | XP_006719957.1 | 4 | 937 | |||
XM_011535309.1 | XP_011533611.1 | 4 | 1186 | |||
XM_017020851.1 | XP_016876340.1 | 4 | 1186 | |||
GenBank mRNA | AF161535.1 | - | 1 | 418 | ||
AJ133352.1 | - | 4 | 715 | |||
AJ133353.1 | - | 4 | 715 | |||
AJ133354.1 | - | 3 | 955 | |||
AJ133355.1 | - | 3 | 955 | |||
BC007048.1 | - | 4 | 699 | 3681047 | ||
BT006687.1 | - | 2 | 562 |