Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DSEL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032160.2 | NP_115536.1 | 2 | 3908 | ||
GenBank mRNA | AF480435.1 | - | 2 | 4160 | ||
AK021539.1 | - | 1 | 2076 | |||
AK021849.1 | - | 2 | 1362 | |||
AK023067.1 | - | 1 | 1728 | |||
BC117325.1 | - | 1 | 2719 | 40125876 |