Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPS36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033281.5 | NP_150597.1 | 3, 4 | 382 | ||
GenBank mRNA | AF086034.1 | - | 2, 3 | 213 | ||
AF271777.1 | - | 486 | ||||
BC015966.1 | - | 3, 4 | 372 | 4068931 | ||
BC017900.1 | - | 3, 4 | 312 | 4251462 | ||
BX951861.1 | - | 3, 4 | 388 |