Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STRADA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003786.2 | NP_001003786.1 | 3, 4 | 399 | ||
NM_001003787.2 | NP_001003787.1 | 5, 6 | 510 | |||
NM_001003788.2 | NP_001003788.1 | 4, 5 | 481 | |||
NM_001165969.1 | NP_001159441.1 | 3, 4 | 423 | |||
NM_001165970.1 | NP_001159442.1 | 3, 4 | 378 | |||
NM_153335.5 | NP_699166.2 | 3, 4 | 399 | |||
XM_005257797.2 | XP_005257854.1 | 5, 6 | 361 | |||
XM_005257798.2 | XP_005257855.1 | 3, 4 | 323 | |||
XM_005257799.2 | XP_005257856.1 | 4, 5 | 332 | |||
XM_005257800.2 | XP_005257857.1 | 5, 6 | 385 | |||
XM_005257801.4 | XP_005257858.1 | 5, 6 | 385 | |||
XM_005257803.4 | XP_005257860.1 | 5, 6 | 385 | |||
XM_011525466.2 | XP_011523768.1 | 5, 6 | 385 | |||
XM_011525467.2 | XP_011523769.1 | 5, 6 | 412 | |||
XM_017025312.1 | XP_016880801.1 | 3, 4 | 274 | |||
XM_017025313.1 | XP_016880802.1 | 4, 5 | 356 | |||
XM_017025314.1 | XP_016880803.1 | 3, 4 | 323 | |||
GenBank mRNA | AF308302.1 | - | 5, 6 | 349 | ||
AK074771.1 | - | 3, 4 | 265 | |||
AK075005.1 | - | 3, 4 | 228 | |||
AK293160.1 | - | 3, 4 | 286 | |||
AK301331.1 | - | 3, 4 | 250 | |||
AL832407.1 | - | 4, 5 | 294 | |||
AY290821.2 | - | 3, 4 | 399 | |||
BC043641.1 | - | 4, 5 | 272 | 3908187 | ||
BG339051.1 | - | 285 |