Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012631.1 | NP_001012649.1 | 8 | 558 | ||
NM_001012632.1 | NP_001012650.1 | 7 | 357 | |||
NM_001012633.1 | NP_001012651.1 | 7 | 491 | |||
NM_001012634.1 | NP_001012652.1 | 6 | 298 | |||
NM_001012635.1 | NP_001012653.1 | 6 | 293 | |||
NM_001012636.1 | NP_001012654.1 | 5 | 281 | |||
NM_001012718.1 | NP_001012736.1 | 7 | 494 | |||
NM_001308078.1 | NP_001295007.1 | 6 | 492 | |||
NM_004221.4 | NP_004212.4 | 7 | 491 | |||
GenBank mRNA | AK296143.1 | - | 6 | 303 | ||
AY495331.1 | - | 7 | 481 | |||
AY495332.1 | - | 7 | 481 | |||
AY495333.1 | - | 6 | 439 | |||
BC009401.2 | - | 6 | 629 | 4300407 | ||
BC018782.1 | - | 7 | 338 | 4873608 | ||
BC105602.1 | - | 7 | 359 | 6496101 | ||
BF568456.1 | - | 6 | 629 | |||
BG748458.1 | - | 7 | 437 | |||
BM759359.1 | - | 4 | 232 | |||
BM822755.1 | - | 8 | 466 | |||
EU339107.1 | - | 4 | 160 | |||
FJ985780.1 | - | 6 | 385 | |||
JN546102.1 | - | 5 | 220 | |||
M59807.1 | - | 6 | 477 |