Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHMP4C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152284.3 | NP_689497.1 | 1, 2 | 358 | ||
GenBank mRNA | AB120734.1 | - | 1, 2 | 190 | ||
AK000049.1 | - | 1, 2 | 321 | |||
AK314873.1 | - | 1, 2 | 357 | |||
AY329086.1 | - | 1, 2 | 192 | |||
BC014321.1 | - | 1, 2 | 362 | 3828410 | ||
BX492886.1 | - | 1, 2 | 378 |