Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MSC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005098.3 | NP_005089.2 | 2 | 883 | ||
GenBank mRNA | AF087036.1 | - | 2 | 551 | ||
AK313102.1 | - | 2 | 840 | |||
BC006313.2 | - | 2 | 849 | 4123825 | ||
BC067827.1 | - | 2 | 858 | 30336424 | ||
BT007169.1 | - | 2 | 565 |