Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MSC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005098.3 | NP_005089.2 | 2 | 915 | ||
GenBank mRNA | AF060154.1 | - | 2 | 808 | ||
AF087036.1 | - | 2 | 583 | |||
AK313102.1 | - | 2 | 872 | |||
BC006313.2 | - | 2 | 881 | 4123825 | ||
BC067827.1 | - | 2 | 890 | 30336424 | ||
BT007169.1 | - | 2 | 597 |