Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MFHAS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004225.2 | NP_004216.2 | 2 | 3623 | ||
GenBank mRNA | AB016816.1 | - | 2 | 3060 | ||
AK027007.1 | - | 2 | 1656 | |||
BC014226.2 | - | 2 | 1914 |