Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYTH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004228.6 | NP_004219.3 | 4 | 558 | ||
NM_017457.5 | NP_059431.1 | 4 | 558 | |||
XM_006723472.2 | XP_006723535.1 | 4 | 644 | |||
XM_006723473.2 | XP_006723536.1 | 4 | 644 | |||
GenBank mRNA | AB209573.1 | - | 4 | 390 | ||
AK292405.1 | - | 4 | 415 | |||
AK299590.1 | - | 4 | 401 | |||
BC004361.1 | - | 4 | 373 | 3538580 | ||
BC038713.1 | - | 4 | 415 | 5269719 | ||
U70728.1 | - | 4 | 416 | |||
X99753.1 | - | 4 | 258 |