Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM114A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138389.2 | NP_612398.2 | 8 | 1126 | ||
NR_033290.1 | 6 | 621 | ||||
XM_005262672.2 | XP_005262729.1 | 7 | 932 | |||
XM_006714033.2 | XP_006714096.1 | 7 | 697 | |||
XM_017008838.1 | XP_016864327.1 | 7 | 932 | |||
GenBank mRNA | AK292831.1 | - | 8 | 1053 | ||
AK296859.1 | - | 4 | 308 | |||
AK304050.1 | - | 6 | 603 | |||
BC001096.2 | - | 6 | 877 | 3507281 | ||
BC040452.1 | - | 8 | 1043 | 3846822 | ||
BX640862.1 | - | 5 | 362 |