Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPR55 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005683.3 | NP_005674.2 | 2 | 483 | ||
XM_005246952.3 | XP_005247009.1 | 2 | 743 | |||
XM_011512175.2 | XP_011510477.1 | 2 | 919 | |||
XM_011512176.2 | XP_011510478.1 | 2 | 502 | |||
GenBank mRNA | AK292223.1 | - | 2 | 628 | ||
BC032694.1 | - | 2 | 483 | 5228678 | ||
CR541776.1 | - | 1 | 290 | |||
FJ348259.1 | - | 1 | 290 |