Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP6V1F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198909.1 | NP_001185838.1 | 1 | 147 | ||
NM_004231.3 | NP_004222.2 | 1 | 147 | |||
GenBank mRNA | AK312214.1 | - | 1 | 92 | ||
BC104230.1 | - | 1 | 98 | 40032234 | ||
BC104231.1 | - | 1 | 98 | 40032235 | ||
BC107854.1 | - | 1 | 124 | 5171984 | ||
BG724326.1 | - | 1 | 132 | |||
BU683155.1 | - | 2 | 598 | |||
BU956696.1 | - | 1 | 46 | |||
CR456896.1 | - | 1 | 45 | |||
D49400.1 | - | 1 | 95 | |||
DB499437.1 | - | 1 | 147 |
Gene Symbol: KCP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK090684.1 | - | 10 | 1409 |