Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DMKN SIRNA Assays ›
Gene Symbol:
DMKN
Gene Name:
dermokine
Gene Aliases:
UNQ729, ZD52F10
Chromosome Location:
Chr.19: 35497217 - 35513678 on Build GRCh38
UniGene ID:
Hs.417795
Species:
Human
siRNA ID
HSS190171
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: DMKN
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001126056.2
NP_001119528.2
2
777
NM_001126057.2
NP_001119529.2
2
777
NM_001126058.2
NP_001119530.2
2
777
NM_001190347.1
NP_001177276.1
2
777
NM_001190348.1
NP_001177277.1
2
777
NM_001190349.1
NP_001177278.1
2
777
NM_033317.4
NP_201574.3
2
777
XM_011527494.2
XP_011525796.1
2
797
XM_011527495.2
XP_011525797.1
2
797
XM_011527496.2
XP_011525798.1
2
797
XM_011527497.2
XP_011525799.1
2
797
XM_011527498.2
XP_011525800.1
2
797
XM_011527499.2
XP_011525801.1
2
797
XM_011527500.2
XP_011525802.1
2
797
XM_011527501.2
XP_011525803.1
2
797
XM_011527502.2
XP_011525804.1
2
797
XM_011527503.2
XP_011525805.1
2
797
XM_011527504.2
XP_011525806.1
2
797
XM_011527505.2
XP_011525807.1
2
797
XM_011527506.2
XP_011525808.1
2
797
XM_011527507.2
XP_011525809.1
2
797
XM_011527508.2
XP_011525810.1
2
797
XM_011527509.2
XP_011525811.1
2
797
XM_011527510.2
XP_011525812.1
2
797
XM_011527511.2
XP_011525813.1
2
797
XM_011527512.2
XP_011525814.1
2
797
GenBank mRNA
AY358412.1
-
2
777
AY622965.1
-
2
594
AY789695.1
-
2
600
AY789696.1
-
2
600
AY789697.1
-
2
600
BC035311.1
-
2
771
4752921
EF158319.1
-
2
600
EF158320.1
-
2
601
EF158322.1
-
2
599
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
cornified envelope assembly
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-65d5d77f49-cp9cp:80/100.66.77.247:80.
git-commit: a4c025c8ba47546866237b65de1f276a5fdbbe78
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.18.1-Offline