Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011655.2 | NP_001011655.1 | 8 | 1205 | ||
NM_001166305.1 | NP_001159777.1 | 9 | 1346 | |||
NM_001166306.1 | NP_001159778.1 | 8 | 1205 | |||
NM_138399.4 | NP_612408.3 | 8 | 1205 | |||
XM_005269371.4 | XP_005269428.1 | 8 | 1219 | |||
XM_011513318.2 | XP_011511620.1 | 9 | 1249 | |||
XM_011513319.2 | XP_011511621.1 | 8 | 1197 | |||
XM_011513320.2 | XP_011511622.1 | 9 | 1427 | |||
XM_011513321.2 | XP_011511623.1 | 8 | 1173 | |||
XM_011513322.2 | XP_011511624.1 | 8 | 1283 | |||
XM_017007517.1 | XP_016863006.1 | 8 | 1107 | |||
XM_017007518.1 | XP_016863007.1 | 8 | 1102 | |||
GenBank mRNA | AK126914.1 | - | 9 | 1002 | ||
AL833026.1 | - | 3 | 763 | |||
AY303778.1 | - | 2 | 195 | |||
AY597813.1 | - | 2 | 195 | |||
BC007772.2 | - | 3 | 351 | |||
BC014883.1 | - | 8 | 1065 | 3907191 | ||
BC034353.1 | - | 3 | 273 | |||
BC111485.1 | - | 9 | 1305 | 5767916 | ||
BC130297.1 | - | 8 | 1109 | 40146328 | ||
BC144158.1 | - | 8 | 1109 | 9052678 | ||
BC144159.1 | - | 8 | 1109 | 9052679 | ||
BX647418.1 | - | 8 | 964 | |||
CF265142.1 | - | 3 | 360 |