Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001314052.1 | NP_001300981.1 | 3 | 1229 | ||
NM_004235.5 | NP_004226.3 | 3 | 1229 | |||
GenBank mRNA | AF022184.1 | - | 3 | 1080 | ||
AF105036.1 | - | 3 | 983 | |||
AK095134.1 | - | 1 | 735 | |||
AK313489.1 | - | 3 | 818 | |||
BC029923.1 | - | 3 | 794 | 5111134 | ||
BC030811.1 | - | 2 | 683 | 4703002 | ||
U70663.1 | - | 3 | 1157 |