Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NREP SIRNA Assays ›
Gene Symbol:
NREP
Gene Name:
neuronal regeneration related protein
Gene Aliases:
C5orf13, D4S114, P311, PRO1873, PTZ17, SEZ17
Chromosome Location:
Chr.5: 111729303 - 111976931 on Build GRCh38
UniGene ID:
Hs.36053
Species:
Human
siRNA ID
HSS190185
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NREP
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001142474.1
NP_001135946.1
4
435
NM_001142475.1
NP_001135947.1
4
465
NM_001142476.1
NP_001135948.1
5
517
NM_001142477.1
NP_001135949.1
5
408
NM_001142478.1
NP_001135950.1
5
384
NM_001142479.1
NP_001135951.1
5
421
NM_001142480.1
NP_001135952.1
5
415
NM_001142481.1
NP_001135953.1
5
324
NM_001142482.1
NP_001135954.1
5
331
NM_001142483.1
NP_001135955.1
3
620
NM_004772.2
NP_004763.1
4
312
XM_006714732.1
XP_006714795.1
3
232
XM_006714733.2
XP_006714796.1
4
213
GenBank mRNA
AF119859.1
-
1
112
AK298779.1
-
4
360
AK315617.1
-
4
286
AL547663.3
-
5
412
BC011050.1
-
5
345
4340005
BC019068.2
-
4
248
4893075
BC072013.1
-
4
271
5922876
BC072443.1
-
4
276
6162335
DA027062.1
-
4
435
DA078027.1
-
3
345
DA520221.1
-
5
343
DA526263.1
-
5
505
DA683910.1
-
5
331
DA694580.1
-
4
330
DA704824.1
-
5
421
DA744980.1
-
5
268
DA752769.1
-
5
324
DA908653.1
-
4
312
DA939543.1
-
5
408
DC365764.1
-
4
465
U30521.1
-
5
343
U36189.1
-
3
156
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
regulation of transforming growth factor beta receptor signaling pathway
axon regeneration
regulation of neuron differentiation
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-65d5d77f49-g5d4h:80/100.66.76.71:80.
git-commit: a4c025c8ba47546866237b65de1f276a5fdbbe78
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.18.1-Offline