Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GTF3C3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012086.4 | NP_036218.1 | 11 | 1574 | ||
XM_005246965.4 | XP_005247022.1 | 11 | 1578 | |||
GenBank mRNA | AF133123.1 | - | 11 | 1510 | ||
AK025560.1 | - | 11 | 1488 | |||
AK027050.1 | - | 5 | 525 | |||
AK314718.1 | - | 11 | 1515 | |||
AL137453.1 | - | 8 | 916 | |||
BC015995.1 | - | 3 | 273 | |||
BC043347.1 | - | 11 | 1429 | 5498609 |