Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SP140L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308162.1 | NP_001295091.1 | 2 | 162 | ||
NM_001308163.1 | NP_001295092.1 | 2 | 162 | |||
NM_138402.4 | NP_612411.4 | 3 | 238 | |||
XM_006712855.2 | XP_006712918.1 | 3 | 224 | |||
XM_006712856.2 | XP_006712919.1 | 3 | 224 | |||
XM_006712858.3 | XP_006712921.1 | 2 | 188 | |||
XM_011512190.2 | XP_011510492.1 | 3 | 224 | |||
XM_011512191.1 | XP_011510493.1 | 3 | 224 | |||
XM_017005294.1 | XP_016860783.1 | 3 | 224 | |||
XM_017005295.1 | XP_016860784.1 | 3 | 224 | |||
XM_017005296.1 | XP_016860785.1 | 3 | 224 | |||
XM_017005297.1 | XP_016860786.1 | 2 | 190 | |||
XM_017005298.1 | XP_016860787.1 | 3 | 224 | |||
XM_017005299.1 | XP_016860788.1 | 2 | 189 | |||
XM_017005300.1 | XP_016860789.1 | 3 | 224 | |||
XM_017005301.1 | XP_016860790.1 | 3 | 224 | |||
GenBank mRNA | BC004921.2 | - | 2 | 144 | ||
DB240458.1 | - | 3 | 224 | |||
DW428200.1 | - | 2 | 180 | |||
KF419365.1 | - | 2 | 72 | |||
KF419366.1 | - | 2 | 72 | |||
KF419367.1 | - | 2 | 72 |