Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNOT8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301073.1 | NP_001288002.1 | 4 | 730 | ||
NM_001301075.1 | NP_001288004.1 | 3 | 584 | |||
NM_001301077.1 | NP_001288006.1 | 4 | 642 | |||
NM_001301080.1 | NP_001288009.1 | 3 | 541 | |||
NM_001301082.1 | NP_001288011.1 | 3 | 254 | |||
NM_001301083.1 | NP_001288012.1 | 2 | 374 | |||
NM_004779.5 | NP_004770.4 | 4 | 778 | |||
XM_011537706.1 | XP_011536008.1 | 5 | 752 | |||
XM_011537708.2 | XP_011536010.1 | 3 | 255 | |||
XM_017010048.1 | XP_016865537.1 | 5 | 637 | |||
XM_017010049.1 | XP_016865538.1 | 5 | 1232 | |||
XM_017010050.1 | XP_016865539.1 | 5 | 1217 | |||
XM_017010051.1 | XP_016865540.1 | 5 | 1267 | |||
XM_017010057.1 | XP_016865546.1 | 5 | 795 | |||
GenBank mRNA | AF053318.1 | - | 4 | 652 | ||
AF087844.1 | - | 3 | 480 | |||
AF180476.1 | - | 3 | 480 | |||
AK025358.1 | - | 4 | 699 | |||
AK297381.1 | - | 3 | 449 | |||
AK299589.1 | - | 5 | 610 | |||
AK309359.1 | - | 4 | 532 | |||
AK314317.1 | - | 4 | 730 | |||
AK315864.1 | - | 3 | 433 | |||
AK316221.1 | - | 3 | 254 | |||
AK316269.1 | - | 2 | 372 | |||
AL122045.1 | - | 4 | 659 | |||
BC017366.2 | - | 4 | 589 | 3506054 | ||
BT006857.1 | - | 3 | 408 | |||
BX647124.1 | - | 2 | 121 | |||
DC387160.1 | - | 4 | 510 |