Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EFTUD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142605.1 | NP_001136077.1 | 16 | 1803 | ||
NM_001258353.1 | NP_001245282.1 | 17 | 2049 | |||
NM_001258354.1 | NP_001245283.1 | 17 | 1882 | |||
NM_004247.3 | NP_004238.3 | 17 | 1912 | |||
GenBank mRNA | AJ505017.1 | - | 13 | 1286 | ||
AK126193.1 | - | 7 | 1379 | |||
AK126464.1 | - | 18 | 1718 | |||
AK126652.1 | - | 2 | 924 | |||
AK128148.1 | - | 17 | 2049 | |||
AK225335.1 | - | 17 | 1672 | |||
AK293108.1 | - | 17 | 1675 | |||
AK296367.1 | - | 16 | 1606 | |||
AK297392.1 | - | 17 | 1688 | |||
AK302828.1 | - | 17 | 1552 | |||
AK316098.1 | - | 16 | 1623 | |||
BC002360.2 | - | 17 | 1682 | 2960986 | ||
CR456774.1 | - | 16 | 1651 | |||
D21163.1 | - | 17 | 1711 |