Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPTE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001141968.1 | NP_001135440.1 | 13 | 1063 | ||
NM_001271850.1 | NP_001258779.1 | 1 | 342 | |||
NM_130785.3 | NP_570141.3 | 12 | 973 | |||
NM_199254.2 | NP_954863.2 | 15 | 1204 | |||
NR_073485.1 | 17 | 1316 | ||||
NR_073486.1 | 10 | 643 | ||||
NR_073487.1 | 9 | 741 | ||||
GenBank mRNA | AJ421032.1 | - | 12 | 973 | ||
AJ421033.1 | - | 9 | 600 | |||
AK301741.1 | - | 17 | 1316 | |||
AY219890.1 | - | 15 | 1204 | |||
BC128146.1 | - | 13 | 1045 | 40112547 | ||
BC128147.1 | - | 9 | 723 | |||
FJ969729.1 | - | 1 | 345 |