Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPL23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000978.3 | NP_000969.1 | 2 | 142 | ||
GenBank mRNA | AK024749.1 | - | 2 | 78 | ||
BC003518.1 | - | 1 | 36 | |||
BC010114.2 | - | 2 | 94 | 3632936 | ||
BC062716.1 | - | 2 | 84 | 4094608 | ||
BC104651.1 | - | 2 | 146 | 6736195 | ||
BC106061.1 | - | 2 | 122 | 4453062 | ||
CA437923.1 | - | 2 | 82 | |||
X52839.1 | - | 2 | 77 | |||
X55954.1 | - | 2 | 64 |
Gene Symbol: SNORA21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK095288.1 | - | 1 | 676 |