Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRMT10A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134665.2 | NP_001128137.1 | 6 | 690 | ||
NM_001134666.2 | NP_001128138.1 | 6 | 679 | |||
NM_152292.4 | NP_689505.1 | 6 | 835 | |||
XM_005263352.3 | XP_005263409.1 | 6 | 669 | |||
XM_006714417.2 | XP_006714480.1 | 6 | 711 | |||
GenBank mRNA | AF106046.1 | - | 4 | 205 | ||
AK000332.1 | - | 6 | 641 | |||
AK313548.1 | - | 6 | 665 | |||
BC028373.2 | - | 6 | 835 | 4838346 | ||
EU668342.1 | - | 6 | 835 |