Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBC1D31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145088.1 | NP_001138560.1 | 13 | 1947 | ||
NM_145647.3 | NP_663622.2 | 13 | 1947 | |||
XM_005251102.1 | XP_005251159.1 | 13 | 1947 | |||
XM_005251103.1 | XP_005251160.1 | 13 | 1947 | |||
XM_005251104.1 | XP_005251161.1 | 13 | 1947 | |||
XM_011517378.2 | XP_011515680.1 | 13 | 2462 | |||
XM_011517379.2 | XP_011515681.1 | 13 | 1947 | |||
XM_017013983.1 | XP_016869472.1 | 13 | 1947 | |||
XM_017013984.1 | XP_016869473.1 | 12 | 2069 | |||
XM_017013985.1 | XP_016869474.1 | 12 | 1942 | |||
XM_017013986.1 | XP_016869475.1 | 12 | 1803 | |||
XM_017013987.1 | XP_016869476.1 | 13 | 1947 | |||
XM_017013988.1 | XP_016869477.1 | 12 | 1782 | |||
XM_017013989.1 | XP_016869478.1 | 12 | 1945 | |||
XM_017013990.1 | XP_016869479.1 | 11 | 1611 | |||
GenBank mRNA | AK025686.1 | - | 7 | 880 | ||
AK056434.1 | - | 13 | 1948 | |||
AK094612.1 | - | 12 | 1745 | |||
BC027237.1 | - | 2 | 219 | |||
BC101724.1 | - | 13 | 1914 | 8069230 | ||
BC111954.1 | - | 13 | 1914 | 8327422 | ||
BC143523.1 | - | 12 | 1798 | |||
BC143525.1 | - | 13 | 1914 | 9052034 |