Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGAP18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033515.2 | NP_277050.2 | 11, 12 | 1667 | ||
XM_005267212.2 | XP_005267269.2 | 11, 12 | 1743 | |||
XM_005267213.1 | XP_005267270.1 | 11, 12 | 1667 | |||
XM_017011494.1 | XP_016866983.1 | 11, 12 | 1743 | |||
GenBank mRNA | AB053293.1 | - | 10, 11 | 1465 | ||
AL834511.1 | - | 11, 12 | 1605 | |||
AY643712.1 | - | 11, 12 | 1668 | |||
BC101708.1 | - | 11, 12 | 1661 | 8069214 | ||
BC107416.1 | - | 11, 12 | 1623 | 4040931 | ||
BC111940.1 | - | 11, 12 | 1661 | 8327408 | ||
BC143531.1 | - | 11, 12 | 1661 |