Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FOXP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172766.2 | NP_001166237.1 | 17 | 2487 | ||
NM_014491.3 | NP_055306.1 | 17 | 2490 | |||
NM_148898.3 | NP_683696.2 | 18 | 2565 | |||
NM_148900.3 | NP_683698.2 | 18 | 2541 | |||
NR_033766.1 | 20 | 2501 | ||||
NR_033767.1 | 18 | 2548 | ||||
XM_017012801.1 | XP_016868290.1 | 20 | 3171 | |||
GenBank mRNA | AF337817.1 | - | 17 | 2266 | ||
AK131266.1 | - | 20 | 2485 | |||
AK296957.1 | - | 18 | 2421 | |||
AY144615.1 | - | 18 | 2560 | |||
BC126104.1 | - | 18 | 2222 | 8991820 | ||
BC143866.1 | - | 18 | 2222 | 9052382 | ||
BC143867.1 | - | 17 | 2161 | 9052383 | ||
DQ778626.1 | - | 12 | 1561 | |||
KJ534839.1 | - | 17 | 2191 | |||
KJ535079.1 | - | 16 | 2329 | |||
KJ535086.1 | - | 16 | 2115 |